missing translation for 'onlineSavingsMsg'
Learn More

Antimony lump, polycrystalline, Puratronic™, 99.9999% (metals basis)

CAS: 7440-36-0 | Sb | 121.76 g/mol

Supplier:  Thermo Scientific Chemicals 01059818

 View more versions of this product

Catalog No. AA1059818


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Antimony lump
50 g
Lump
Sb
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
Puratronic
7440-36-0
Odorless
(metals basis)
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
distibyne
5354495
99.9999%

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
[Sb]
Sb
MFCD00011211 MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-146-5

RTECSNumber : CC4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only