missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 99.999% (metals basis)

Catalog No. p-7048859
Change view
Click to view available options
Quantity:
10 g
50 g
Catalog No. Quantity
AA1168309 10 g
AA1168318 50 g
2 options

Catalog No. AA1168309

Supplier: Thermo Scientific Chemicals 1168309

Only null left

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

It is used in glitter compositions, fountain compositions, flash powder and vulcanizing rubber. It is employed in the production of ruby-colored glass and in plastics, as a fire retardant, in explosives, matches, and fireworks. It can be used to prepare a series of thioantimonous salts and Schlippe's salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Physical Form Powder
Quantity 10 g
UN Number UN1549
Merck Index 14,713
Solubility Information Insoluble in water,and moderately soluble in acids.
Formula Weight 339.68
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Antimony(III) sulfide
Hazard Category H302-H312-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber 215-713-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only