missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 99.999% (metals basis)

Catalog No. AA4546409
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA4546409 10 g Glass bottle
AA4546418 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4546409 Supplier Thermo Scientific Chemicals Supplier No. 04546409

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

Antimony(III) sulfide is manufactured for use in safety matches, military ammunition, explosives and fireworks. Also it is used in the production of ruby-colored glass and in plastics as a flame retardant. It is a semiconductor with a direct band gap of 1.8-2.5 eV. With suitable doping p and n type materials can be produced.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Physical Form Pieces
Quantity 10 g
UN Number UN1549
Solubility Information Insoluble in water.
Formula Weight 339.68
Percent Purity 99.999%
Packaging Glass bottle
Chemical Name or Material Antimony(III) sulfide
Hazard Category H302-H312-H315-H319-H335-H500
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 215-713-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only