missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 99.9% (metals basis)

Catalog No. AA4037109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4037109 10 g
AA4037118 50 g
AA4037130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4037109 Supplier Thermo Scientific Chemicals Supplier No. 04037109

Vacuum deposition | CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

Antimony Trisulfide is used in the production of pyrotechnics and as a yellow pigment for glass and porcelain. In Vacuum deposition. Used in preparation of Vermilion or yellow pigment, for antimony salts, percussion caps, camouflage paints, and in ruby glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Physical Form Pieces
Quantity 10 g
UN Number UN1549
Merck Index 14,713
Solubility Information Insoluble in water. Soluble in concentrated acids.
Formula Weight 339.68
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Antimony(III) sulfide
Hazard Category H302-H312-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber 215-713-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only