missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 98%

Catalog No. AC223591000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223591000 100 g Glass bottle
AC223595000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223591000 Supplier Thermo Scientific Chemicals Supplier No. 223591000

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
Synonym Antimony trisulfide
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1150°C
Melting Point 550°C
Color Gray to Black
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 97.5% min. (Sb2S3)
Linear Formula Sb2S3
Merck Index 15, 701
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. hcl and alkali hydroxides,soluble in alcohol,soluble in amminium hydrogen sulfide,insoluble in acetic acid
Formula Weight 339.67
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Antimony(III) sulfide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 215-713-4
RTECSNumber CC9450000
TSCA TSCA

RUO – Research Use Only