missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) isopropoxide, 99.9% (metals basis)

Catalog No. AA3658914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3658914 25 g
AA3658906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3658914 Supplier Thermo Scientific Chemicals Supplier No. 03658914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18770-47-3 | C9H21O3Sb | 299.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18770-47-3
Molecular Formula C9H21O3Sb
Molecular Weight (g/mol) 299.024
MDL Number MFCD00143623
InChI Key HYPTXUAFIRUIRD-UHFFFAOYSA-N
Synonym antimony iii isopropoxide, antimony 3+ tripropan-2-olate, triisopropoxyantimony, acmc-209epx, antimony iii propan-2-olate, antimony 3+ ; propan-2-olate, 2-propanol,antimony 3+ salt 9ci, antimony 3+ ion tris propan-2-olate, antimony 3+ tris propan-2-olate
PubChem CID 3034303
IUPAC Name antimony(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Sb+3]

Specifications

Color Colorless
Density 1.31 g/mL
Boiling Point 68°C (3 mmHg)
Flash Point 77°C (141°F)
Quantity 25 g
UN Number UN3082
Sensitivity Moisture sensitive
Formula Weight 298.8
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Antimony(III) isopropoxide
Show More Show Less
Hazard Category H227-H302+H332
Hazard Statement GHS H Statement
H302-H332-H227
Harmful if swallowed.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 242-561-6
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only