missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) ethoxide, 99.9% (metals basis)

Catalog No. AA4309706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4309706 5 g
AA4309714 25 g
AA4309722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4309706 Supplier Thermo Scientific Chemicals Supplier No. 04309706

CAS: 10433-06-4 | C6H15O3Sb | 256.943 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10433-06-4
Molecular Formula C6H15O3Sb
Molecular Weight (g/mol) 256.943
MDL Number MFCD00015123
InChI Key JGOJQVLHSPGMOC-UHFFFAOYSA-N
Synonym antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide
PubChem CID 66326
IUPAC Name antimony(3+);ethanolate
SMILES CC[O-].CC[O-].CC[O-].[Sb+3]

Specifications

Density 1.524 g/mL
Boiling Point 95°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3082
Sensitivity Moisture sensitive
Formula Weight 256.9
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Antimony (III) ethoxide
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 233-914-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only