missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) Chloride, ACS, For Analysis, EMSURE™, MilliporeSigma™
$367.58 - $922.14
Chemical Identifiers
| CAS | 10025-91-9 |
|---|---|
| Molecular Formula | Cl3Sb |
| Molecular Weight (g/mol) | 228.11 |
| MDL Number | MFCD00011212 |
| InChI Key | FAPDDOBMIUGHIN-UHFFFAOYSA-K |
| Synonym | antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony |
| PubChem CID | 24814 |
| ChEBI | CHEBI:74856 |
| IUPAC Name | antimony(3+) trichloride |
| SMILES | [Cl-].[Cl-].[Cl-].[Sb+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1078380250
|
Technidata
1078380250 |
250 g | Glass bottle |
Each for $367.58
|
|
||||
|
M1078381000
|
Technidata
1078381000 |
1 kg | Glass bottle |
Each for $922.14
|
|
||||
Description
For analysisChemical Identifiers
| 10025-91-9 | |
| 228.11 | |
| FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
| 24814 | |
| antimony(3+) trichloride |
| Cl3Sb | |
| MFCD00011212 | |
| antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
| CHEBI:74856 | |
| [Cl-].[Cl-].[Cl-].[Sb+3] |
Specifications
| 10025-91-9 | |
| 250 g | |
| MFCD00011212 | |
| antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
| [Cl-].[Cl-].[Cl-].[Sb+3] | |
| 228.11 | |
| CHEBI:74856 | |
| ACS | |
| Glass bottle | |
| 0.16 hPa at 20°C |
| Solid | |
| Cl3Sb | |
| UN 1733 | |
| FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
| antimony(3+) trichloride | |
| 24814 | |
| 228.1 | |
| Stinging | |
| Colorless solid | |
| Antimony(III) chloride |
Safety and Handling
H314: Causes severe skin burns and eye damage.
H411: Toxic to aquatic life with long lasting effects.
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 233-047-2