missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) Chloride, ACS, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1078380250
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1078380250 250 g Glass bottle
M1078381000 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1078380250 Supplier Technidata Supplier No. 1.07838.0250
For analysis

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name antimony(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Physical Form Solid
Quantity 250 g
UN Number UN 1733
Formula Weight 228.1
Grade ACS
Odor Stinging
Packaging Glass bottle
Appearance Colorless solid
Vapor Pressure 0.16 hPa at 20°C
Chemical Name or Material Antimony(III) chloride
Hazard Category H314: Causes severe skin burns and eye damage.
H411: Toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 233-047-2