missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, ACS, 99.0% min

Catalog No. p-4407245
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3628118 50 g
AA3628130 250 g
AA36281A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3628118 Supplier Thermo Scientific Chemicals Supplier No. 03628118
Only null left

CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol

Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols and carotenoids. It is likewise employed as a dye mordant and a fabric flame retardant. It is used as inorganic and organic chlorination catalyst. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99% min.
Solubility Information Soluble in water,HCl solution,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone,carbon tetrachloride. Insoluble in organic bases.
Formula Weight 228.11
Grade ACS
Odor Pungent
Sensitivity Moisture sensitive
Appearance White crystalline
Density 3.14 g/cm3
Chemical Name or Material Antimony(III) chloride, For analysis ACS
Show More Show Less
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 233-047-2
RTECSNumber CC4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only