missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, 99.9% (metals basis)

Catalog No. AA4153714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4153714 25 g
AA4153722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4153714 Supplier Thermo Scientific Chemicals Supplier No. 04153714

CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol

Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water,hydrochloric acid,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone and carbon tetrachloride.
Formula Weight 228.11
Odor Pungent
Sensitivity Moisture sensitive
Density 3.14
Chemical Name or Material Antimony(III) chloride
Show More Show Less
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 233-047-2
RTECSNumber CC4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only