missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) bromide, 99%

Catalog No. AA4157318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4157318 50 g
AA4157330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4157318 Supplier Thermo Scientific Chemicals Supplier No. 04157318

CAS: 7789-61-9 | Br3Sb | 361.472 g/mol

Antimony(III) bromide is used in the synthesis of antimony compounds, in semiconductors and chemical analysis. It acts as a fire retardant in polyethylene polymers, as mordant and in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.472
MDL Number MFCD00016317
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br

Specifications

Boiling Point 280°C
Melting Point 96.6°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%
Solubility Information Soluble in alcohol,acetone,ammonia,benzene,chloroform,dilute hydrochloric acid,hydrobromic acid and carbon disulfide.
Formula Weight 361.48
Odor Characteristic
Refractive Index 1.74
Sensitivity Moisture sensitive
Appearance White powder
Density 4.148 g/cm3
Chemical Name or Material Antimony(III) bromide
Show More Show Less
Hazard Category H302+H332-H335
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation DOT Class: 6.1; Packaging Group: III
EINECSNumber 232-179-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only