missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) bromide, 99.995% (metals basis), Thermo Scientific Chemicals

$233.01 - $233.01

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.472
MDL Number MFCD00016317
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1168609
View Documents
Thermo Scientific Chemicals
01168609
10 g
Each for $233.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-61-9
361.472
RPJGYLSSECYURW-UHFFFAOYSA-K
24615
Br[Sb](Br)Br
Br3Sb
MFCD00016317
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
tribromostibane

Specifications

280°C
7789-61-9
10 g
MFCD00016317
14,706
RPJGYLSSECYURW-UHFFFAOYSA-K
tribromostibane
24615
99.995%
1.74
4.148 g/mL
(metals basis)
96.6°C
Crystalline
Br3Sb
UN3260
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
Br[Sb](Br)Br
361.472
361.48
Characteristic
Moisture sensitive
Antimony(III) bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 232-179-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only