missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) bromide, 99.5% (metals basis)

Catalog No. p-7051578
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4157409 10 g
AA4157418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4157409 Supplier Thermo Scientific Chemicals Supplier No. 04157409
Only null left

CAS: 7789-61-9 | Br3Sb | 361.472 g/mol

Antimony(III) bromide is used in the production of other antimony compounds, in chemical analysis, as a mordant, and in dyeing. It can be added to polymers such as polyethylene as a fire retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.472
MDL Number MFCD00016317
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br

Specifications

Boiling Point 280°C
Melting Point 96.6°C
Quantity 10 g
UN Number UN3260
Merck Index 14,706
Solubility Information Soluble in dilute hydrochloric acid,hydrobromic acid,carbon disulfide,alcohol,acetone,ammonia,benzene,chloroform.
Formula Weight 361.48
Percent Purity 99.5%
Refractive Index 1.74
Sensitivity Moisture sensitive
Density 4.148 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(III) bromide
Show More Show Less
Hazard Category H302+H332-H335
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 232-179-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only