missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) bromide, 99.5% (metals basis)
Description
Antimony(III) bromide is used in the production of other antimony compounds, in chemical analysis, as a mordant, and in dyeing. It can be added to polymers such as polyethylene as a fire retardant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 280°C |
| Melting Point | 96.6°C |
| Quantity | 10 g |
| Refractive Index | 1.74 |
| Sensitivity | Moisture sensitive |
| Density | 4.148 g/mL |
| UN Number | UN3260 |
| Merck Index | 14,706 |
| Solubility Information | Soluble in dilute hydrochloric acid,hydrobromic acid,carbon disulfide,alcohol,acetone,ammonia,benzene,chloroform. |
| Formula Weight | 361.48 |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H335 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 232-179-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only