missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antifoam AF, Emulsion, Food Grade, Spectrum™ Chemical
Description
Spectrum™ Chemical Antifoam AF, Emulsion, Food Grade is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 100 mL |
| Grade | Food Grade |
| Packaging | Poly Bottle |