missing translation for 'onlineSavingsMsg'
Learn More

Antifoam AF, Emulsion, Food Grade, Spectrum™ Chemical

Catalog No. 18600728
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Cubitainer
Drum
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600728 100 mL Poly Bottle
18600729 500 mL Poly Bottle
18600730 4 L Poly Bottle
18600732 20 L Cubitainer
18600731 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600728 Supplier Spectrum Chemical Mfg Cor Supplier No. A1303100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

A1303

Spectrum™ Chemical Antifoam AF, Emulsion, Food Grade is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 67762-90-7

Specifications

CAS Max % 100%
Quantity 100 mL
Grade Food Grade
Packaging Poly Bottle