Learn More
Contec™ Anticon™ White Magic™ Wipes
GREENER_CHOICE
Anticon™ White Magic™ Wipes are made from 100% continuous filament polyester with an interlock construction and excellent sorbency capacity. Exhibit low particle and fiber counts and provides ultra-clean wiping for critical applications.
$1,283.07
Specifications
Product Type Wipe
Material Polyester Knit
Disposable Single-use
Edges Laser cut
For Use With (Application) Process tools and other equipment
View More Specs
Products 1
missing translation for 'productCode' missing translation for 'brand' Dimensions (L x W) Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Dimensions (L x W) Price Quantity & Availability  
19168534
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Contec™
495353323
12 x 12 in.
Case of 800 Each for $1,283.07
Only null left
Description
Description
  • Low sub-micron particles and fiber count
  • Patented sorbency technology
  • Gamma irradiated to a Sterility Assurance Level of 10-6 SAL
  • Minimizes the risk of contamination
  • Reduces the number of wipes required
  • Ideal for applications that require high sorbency
  • Eliminates the introduction of biological contamination
Specifications
Specifications
Wipe
Single-use
Process tools and other equipment
Polyester Knit
Laser cut
Sterile