This ready-to-use Antibody Dilution Buffer is designed for the dilution of antibodies in all immunoassays, including immunofluorescence, IHC, ELISA, WB. This buffer contains immunoglobulin free BSA and does not contain phosphate, sodium azide, or mercury preservatives. It can be used for the dilution of all antibodies, including peroxidase, and antibodies to phophoproteins. However, it is not suitable for dilution of antibodies to S100 proteins. This buffer contains green food color.
Application: Dilute antibodies, primary or secondary as recommended in the protocol by the manufacturer of these products.
Storage: 2 8 degree C. Diluted antibodies can be stored at 2 8 degree C for several weeks, depending upon the stability of antibodies.