Learn More
Impacto™ Anti-Vibration Half Finger Pearl Leather AirGloves™
Catalog No. 19400058
Encompass
Change view
Click to view available options
Size:
2X-Large
Catalog No. Size
19400058 2X-Large
1 options
Catalog No. 19400058 Supplier Impacto™ Supplier No. BG40160
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Gloves utilize patented technology in the palm, fingers, and thumb for protection against impact and vibration.
  • Gloves offer comfortable hand protection from impact and vibration
  • Designed to help prevent Hand-Arm Vibration syndrome (HAVS)
  • Patented air bladder technology protects the palm, fingers and thumb from tool-induced vibration
  • Thin urethane air bladder ensures tactile feedback to minimize grip stress
  • Soft leather palm and thumb for tactile feedback and dexterity
  • Opposed thumb helps maintain vibration protection when clasping tool handles
  • Individual chambers for long lasting wear and consistent protection
  • Adjustable hook and loop closure at the wrist
Specifications
Back Material Nylon
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Cuff Material Elastic
Color Black, White
Hand Specific
Material Pearl Leather
Size 2X-Large
Finger Style Half
Glove Cuff Style Hook and Loop
Series BG401