Learn More
Innovatek Anti-Sterptolysin O (ASO) Latex Test Kits
Description
- Fast 2 to 3 minute rotation
- Complete kit includes all reagents and controls
- Disposable test cards
- 6 to 12 months of shelf life
- Sensitive and specific
Specifications
Specifications
| Detectable Analytes | Anti-Streptolysin O (ASO) |
| Disposable | Single-use |
| Quantity | 50 Tests/Kit |
| Test Time | ≈3 min. |