missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KNF™ FLEXPAK Anti-static Block Bottom Bags
Clear and clean polyethylene bags with a 4 mil thickness.
Supplier: KNF™ FLEXPAK 020507287
Description
- Clear, anti-static plastic
- Square bottom
Chemical Identifiers
| Not Autoclavable | |
| Clear | |
| 4 mil |
| Level 200 Clean | |
| Polyethylene | |
| 0.1016 mm |
Specifications
| Not Autoclavable | |
| Clear | |
| 4 mil | |
| 101.6 cm | |
| 71.1 cm |
| Level 200 Clean | |
| Polyethylene | |
| 40 x 28 in. | |
| 101.6 cm | |
| 0.1016 mm |