Learn More
KNF™ FLEXPAK Anti-static Block Bottom Bags
Clear and clean polyethylene bags with a 4 mil thickness.
Supplier:  KNF™ FLEXPAK 020507244
 View more versions of this product
Catalog No. 17020170
Description
Description
  • Clear, anti-static plastic
  • Square bottom
Specifications
Chemical Identifiers
Not Autoclavable
Clear
4 mil
Level 200 Clean
Polyethylene
0.1016 mm
Specifications
Not Autoclavable
Clear
4 mil
81.3 cm
81.9 cm
Level 200 Clean
Polyethylene
32.25 x 32.25 in.
81.9 cm
0.1016 mm
Product Suggestions
Product Suggestions