Learn More
SOMNI Scientific™ Anti-Spil Isoflurane Bottle Adapter
Catalog No. 04777189
Encompass
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04777189 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04777189 Supplier SOMNI Scientific™ Supplier No. VF6230
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Allows for safe transfer of anesthetic from bottle to vaporizer. The curved tip and metered flow reduce the risk of spilling while the antipollution plug prevents agent loss during storage.
  • The Anti-Spil Isoflurane Bottle Adapter allows for safe transfer of anesthetic from bottle to vaporizer
  • Curved tip and metered flow reduce the risk of spilling
  • The antipollution plug prevents agent loss during storage
  • Reduce exposure to Isoflurane
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Type Bottle Adapter