Learn More
ACROBiosystems Inc ACROBiosystems Anti-Rituximab Antibodies (MALS verified)
Catalog No. 502106932
Encompass
Antibody;1MG;IgG1 / kappa;Recognizes Rituximab specifically, no cross reactivity with other humanized antibodies.;Anti-Rituximab Antibodies (RIB-Y35c) are expressed from human 293 cells (HEK293).;This protein . ;Immobilized Rituximab at 1 g/mL (100 L/well) can bind Anti-Rituximab Antibodies (Cat. No. RIB-Y35c) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 502106932 Supplier ACROBiosystems Inc Supplier No. RIBY35C1MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Antibody;1MG;IgG1 / kappa;Recognizes Rituximab specifically, no cross reactivity with other humanized antibodies.;Anti-Rituximab Antibodies (RIB-Y35c) are expressed from human 293 cells (HEK293).;This protein . ;Immobilized Rituximab at 1 g/mL (100 L/well) can bind Anti-Rituximab Antibodies (Cat. No. RIB-Y35c) with a linear range of 0.2-6 ng/mL (QC tested).