missing translation for 'onlineSavingsMsg'
Learn More

anti-PEN2 Polyclonal, Invitrogen™

PEN2 Polyclonal antibody PA1-10293 from Thermo Scientific Pierce specifically detects PEN2 in Hamster and Mouse samples, and it is validated for WB

Supplier:  METTLER TOLEDO 30601008

Catalog No. 01-670-249


Only null left
Explore available promotions

Description

Description

Presenilins, which are components of the gamma-secretase protein complex, are required for intramembranous processing of some type I transmembrane proteins, such as the Notch proteins and the beta-amyloid precursor protein. Signaling by Notch receptors mediates a wide range of developmental cell fates. Processing of the beta-amyloid precursor protein generates neurotoxic amyloid beta peptides, the major component of senile plaques associated with Alzheimer's disease. This gene encodes a protein that is required for Notch pathway signaling, and for the activity and accumulation of gamma-secretase.

Host Species: Rabbit
Isotype: IgG
Species Reactivity: Hamster, Human, Mouse
Formulation: 200μg of Affinity chromatography purified antibody ;
Immunogen: Synthetic peptide, conjugated to KLH, corresponding to the 11 most N-terminal residues of human Pen-2.
Synonyms: hPen2-N
Uniprot Accession: Q9NZ42, Q9CQR7
Entrez Gene ID: 66340, 55851

PA1-10293 detects PEN2 in Hamster and Mouse samples. PA1-10293 has been successfully used in WB procedures. The immunogen is Synthetic peptide, conjugated to KLH, corresponding to the 11 most N-terminal residues of human Pen-2.

Western Blotting

Specifications

Specifications

PEN2
Unconjugated
Synthetic peptide, conjugated to KLH, corresponding to the 11 most N-terminal residues of human Pen-2.
Primary
Store at -20°C. Avoid Freeze/Thaw Cycles
IgG
Polyclonal
Rabbit
200 μg
Hamster, Human, Murine
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions