Learn More
Enzo Life Sciences NLRP3/NALP3 (human) monoclonal antibody (Nalpy3-b) (100 g) DFS Item
Supplier:  Enzo ALX804819C100
Clone: Nalpy3-b. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human NLRP3/NALP3 (NACHT-, LRR- and PYD-containing protein 3)(pyrin domain). UniProt: Q96P20. Source: Purified from concentrated hybridoma tissue culture supernatant. Species Reactivity: Human. Specificity: Detects endogenous protein by IP and WB. Applications: IHC (FS), IP, WB. Recommended dilutions: Immunoprecipitation (1:50). Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. NC9771950
Only null left
Explore available promotions