Learn More
CD7/C13, FITC, PE, clone: MG34, E735, Invitrogen™
Antibody Cocktail
Supplier: Bio-Serv BIO9170
Description
CD7 is a 40kD transmembrane, single-chain glycoprotein. CD7 is expressed by majority of peripheral blood T cells, NK cells and all Thymocytes. The presence or absence of CD7 is a useful marker for the classification of T- and NK-cell malignancies. The CD7 molecule is absent in a subset of human T cells under certain physiologic conditions. Thus an increase in CD7 negative T cells can be seen in some inflammatory skin disease. This antibody recognizes the human CD13 cell surface glycoprotein, a 150kD molecule expressed by granulocytes and monocytes, and by myeloid leukemia cells. Inhibits infection of cells by human coronavirus, inhibits aminopeptidase N activity of the CD13 molecule immunoprecipitates. MA1-22542 detects CD13 in Human samples.
This antibody is a monoclonal mixture of FITC-conjugated CD7 (clone MG34, isotype IgG2a) and PE-conjugated CD13 (clone E735, isotype IgG1).Specifications
| CD7 + C13 | |
| Cocktail | |
| FITC/PE | |
| ANPEP | |
| GP40, LEU-9, TP41, Tp40, ANPEP, AP-M, AP-N, Apn, Cd13, P150, alanyl aminopeptidase, aminopeptidase M, aminopeptidase N, aminopeptidase N/CD13, membrane protein p161, microsomal aminopeptidase | |
| Murine | |
| Protein A/G, Protein A/G | |
| RUO | |
| 290, 290, 924, 924 | |
| Store at 4°C, do not freeze | |
| Purified |
| Flow Cytometry | |
| MG34, E735, E735 | |
| PBS with 0.2% carrier protein and 0.08% sodium azide | |
| P09564, P15144 | |
| ANPEP, CD7 | |
| T-acute lymphoblastic leukemia (T-ALL) cells | |
| 50 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1, IgG2a |