missing translation for 'onlineSavingsMsg'
Learn More

anti-(1R)-(+)-Camphorquinone 3-Oxime 95.0+%, TCI America™

Catalog No. C16611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16611G Supplier TCI America Supplier No. C16611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31571-14-9
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD00074848
InChI Key YRNPDSREMSMKIY-WKEGUHRASA-N
Synonym anti-1r-+-camphorquinone3-oxime, 1s,4r-2-hydroxyimino-4,7,7-trimethylbicyclo 2.2.1 heptan-3-one
PubChem CID 23308125
IUPAC Name (1R,4R)-2-hydroxyimino-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC1(C(=O)C2=NO)C)C

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 1 g
Formula Weight 181.24
Percent Purity ≥95.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material anti-(1R)-(+)-Camphorquinone 3-Oxime