missing translation for 'onlineSavingsMsg'
Learn More

Anthrarufin 85.0+%, TCI America™

Catalog No. D056225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D056225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D056225G Supplier TCI America Supplier No. D056225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 117-12-4
Melting Point 280°C
Molecular Formula C14H8O4
Quantity 25 g
MDL Number MFCD00001210
Synonym anthrarufin, 1,5-dihydroxyanthraquinone, 1,5-dihydroxyanthrachinon, 9,10-anthracenedione, 1,5-dihydroxy, 1,5-dihydroxy-9,10-anthraquinone, 1,5-dihydroxy-9,10-anthracenedione, anthraquinone, 1,5-dihydroxy, az-u, unii-kpb60w5s3m, ccris 3150
InChI Key JPICKYUTICNNNJ-UHFFFAOYSA-N
SMILES OC1=CC=CC2=C1C(=O)C1=CC=CC(O)=C1C2=O
IUPAC Name 1,5-dihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular Weight (g/mol) 240.21
PubChem CID 8328
ChEBI CHEBI:37501
Formula Weight 240.21
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Anthrarufin
Show More Show Less
EINECSNumber (4)-0704
RTECSNumber CB6630000
TSCA Yes