missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone 98.0+%, TCI America™

Catalog No. A050225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A050225G 25 g
A0502500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A050225G Supplier TCI America Supplier No. A050225G

Chemical Identifiers

CAS 84-65-1
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.22
MDL Number MFCD00001188
InChI Key RZVHIXYEVGDQDX-UHFFFAOYSA-N
Synonym anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone
PubChem CID 6780
ChEBI CHEBI:40448
IUPAC Name 9,10-dihydroanthracene-9,10-dione
SMILES O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12

Specifications

Melting Point 284°C
Boiling Point 380°C
Quantity 25 g
UN Number 3077
Formula Weight 208.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Anthraquinone
EINECSNumber (4)-0686
RTECSNumber CB4725000
TSCA Yes