missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone-1,5-disulfonic Acid Disodium Salt 98.0+%, TCI America™

Catalog No. a050625g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A050625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A050625G Supplier TCI America Supplier No. A050625G

Specifications

CAS 853-35-0
Molecular Formula C14H6Na2O8S2
Quantity 25 g
MDL Number MFCD00044659
Synonym anthraquinone-1,5-disulfonic acid disodium salt, 1,5-anthraquinonedisulfonic acid, disodium salt, sodium anthraquinone-1,5-disulfonate, disodium anthraquinone-1,5-disulfonate, 1,5-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, disodium salt, anthrachinon-1,5-disulfonan sodny czech, anthrachinon-1,5-disulfonan disodny czech, disodium 9,10-dihydro-9,10-dioxoanthracene-1,5-disulphonate, 1,5-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt 1:2, 1,5-bis sodiooxysulfonyl-9,10-anthraquinone
InChI Key OFASSSMJNCWWTP-UHFFFAOYSA-L
SMILES C1=CC2=C(C(=C1)S(=O)(=O)[O-])C(=O)C3=C(C2=O)C(=CC=C3)S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;9,10-dioxoanthracene-1,5-disulfonate
Molecular Weight (g/mol) 412.294
PubChem CID 70067
Formula Weight 412.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Anthraquinone-1,5-disulfonic Acid Disodium Salt
Show More Show Less
EINECSNumber (4)-0725
RTECSNumber CB0555500
TSCA Yes