missing translation for 'onlineSavingsMsg'
Learn More

Anthraniloyl Hydrazine 98.0+%, TCI America™

Catalog No. A110010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A110010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A110010G Supplier TCI America Supplier No. A110010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1904-58-1
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017056
InChI Key GRWMSCBKWMQPON-UHFFFAOYSA-N
Synonym 2-aminobenzhydrazide, o-aminobenzhydrazide, anthranilic acid hydrazide, benzoic acid, 2-amino-, hydrazide, anthranilic hydrazide, anthraniloyl hydrazide, anthraniloyl hydrazine, 2-aminobenzoylhydrazine, 2-aminobenzoic acid hydrazide, o-aminobenzoic hydrazide
PubChem CID 94728
IUPAC Name 2-aminobenzohydrazide
SMILES NNC(=O)C1=CC=CC=C1N

Specifications

Melting Point 123°C
Quantity 10 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Anthraniloyl Hydrazine