missing translation for 'onlineSavingsMsg'
Learn More

Ansul™ Dry Chemicals

Catalog No. p-38634
Change view
Click to view available options
Product Type:
Foray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
296359800 Foray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296359800 Supplier Ansul™ Supplier No. 53080
Only null left

Dry chemicals are available to fight Class A, B, C, or D fires

Never mix bicarbonate-based and phosphate-based dry chemicals. A chemical reaction that is harmful to the extinguisher will occur. Foray™ is free flowing, water repellent, nonabrasive; and produces no toxic effects. Other additives to Met-L-X™ render it free-flowing and cause heat caking or crusting.

Foray

  • For Class A, B or C fires
  • Composed of a monoammonium phosphate base
  • Useful on combination fires and on Class A fires at temperatures that freeze water; applications include heavy equipment used in the logging industry
  • May produce mild corrosion after fire, making prompt cleanup necessary; if agent cannot be readily swept up, warm water may be used as a solvent

Met-L-X

  • For Class D (sodium, potassium, sodium-potassium alloy or magnesium) fires
  • Composed of a salt base plus a polymer for sealing
  • Controls and sometimes extinguishes small fires on zirconium and titanium
  • UL listed

Specifications

Product Type Foray
Formulation Composed of a salt base plus a polymer for sealing
For Use With (Application) Class A, B, C fires
Weight (English) 45 lb.
Weight (Metric) 20.4 kg