missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ANSI Sign: TRIPPING HAZARD. FALLS MAY RESULT IN INJURY.

Catalog No. 19112866
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
Material:
Fiberglass
Polystyrene
Max. Temperature (English):
176°F
194°F
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material Max. Temperature (English)
19112866 14 x 10 in. Fiberglass 194°F
19112869 10 x 7 in. Polystyrene 176°F
19112870 14 x 10 in. Polystyrene 176°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19112866 Supplier Brady™ Supplier No. 144763

Improve workplace safety with non-adhesive mounted signage. The Brady™ Tripping Hazard ANSI Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

  • Abrasion-resistant and chemical-resistant
  • Corner holes provide easy mounting solutions in a variety of environments

Specifications

Finish Not Laminated
Material Fiberglass
Quantity 1 Ea.
Mounting Type Corner Holes
Height (English) 10 in.
Max. Temperature (English) 194°F
Min. Temperature (English) -40°F
Thickness 0.0984 in.
Width (English) 14 in.
Shape Rectangular
Temperature Range (English) -40°F to 194°F
Dimensions (L x W) 14 x 10 in.
Displayed Text TRIPPING HAZARD. FALLS MAY RESULT IN INJURY.
Color Yellow on White
Show More Show Less