missing translation for 'onlineSavingsMsg'
Learn More

Ansell Touch N Tuff™ Disposable Nitrile Gloves

Catalog No. 18999266d
Change view
Click to view available options
:
Lightly powdered; 9.5 in. long (24cm); Size: X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18999266D Lightly powdered; 9.5 in. long (24cm); Size: X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999266D Supplier Ansell Supplier No. 9250011100
Only null left

Proven splash resistance against hazardous chemicals. Ansell Edmont Touch N Tuff™ Disposable Nitrile Gloves provide a comfortable fit and minimize hand fatigue.

Proven splash resistance against hazardous chemicals. Ansell Edmont Touch N Tuff™ Disposable Nitrile Gloves provide a comfortable fit and minimize hand fatigue.

Offers exceptional resistance to nuisance punctures, nicks, cuts and abrasions. Splash protection against irritating chemicals.

  • Made from 100% nitrile for superior elasticity
  • Comfortable thin mil construction provides good chemical resistance
  • Rolled cuffs for easy donning and doffing
  • Green
Not recommended for Acetone, Benzaldehyde, Chlorobenzene, EDC, DMAC, DMF, Ethyl Acetate, Furfural, Methylene Bromide, MEK, Morpholine, NMP, Iodomethane, Nitric Acid -70%, (Concentrated), itric Acid, Red Fuming, Nitrobenzene, 1-Nitropropane, 2-Nitropropane, Phenol, 90%, THF, Pyridine, Sulfuric Acid, 95-98% (Concentrated) and THF

Specifications

Certifications/Compliance FDA 21 CFR 177-2600
Color Green
Disposable Single-use
Material Nitrile
Powdered Powdered
Size X-Large
Thickness (English) 5 mil
Thickness (Metric) 5 mm
Model 92 500 XL585715
Tensile Strength 14 MPa
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less