missing translation for 'onlineSavingsMsg'
Learn More

Ansell Touch N Tuff™ Disposable Nitrile Gloves

Catalog No. 18999266C
Change view
Click to view available options
Tensile Strength:
14 MPa
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tensile Strength
18999266C 14 MPa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999266C Supplier Ansell Supplier No. 9250011090
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Proven splash resistance against hazardous chemicals. Ansell Edmont Touch N Tuff™ Disposable Nitrile Gloves provide a comfortable fit and minimize hand fatigue.

Proven splash resistance against hazardous chemicals. Ansell Edmont Touch N Tuff™ Disposable Nitrile Gloves provide a comfortable fit and minimize hand fatigue.

Offers exceptional resistance to nuisance punctures, nicks, cuts and abrasions. Splash protection against irritating chemicals.

  • Made from 100% nitrile for superior elasticity
  • Comfortable thin mil construction provides good chemical resistance
  • Rolled cuffs for easy donning and doffing
  • Green
Not recommended for Acetone, Benzaldehyde, Chlorobenzene, EDC, DMAC, DMF, Ethyl Acetate, Furfural, Methylene Bromide, MEK, Morpholine, NMP, Iodomethane, Nitric Acid -70%, (Concentrated), itric Acid, Red Fuming, Nitrobenzene, 1-Nitropropane, 2-Nitropropane, Phenol, 90%, THF, Pyridine, Sulfuric Acid, 95-98% (Concentrated) and THF

Specifications

Certifications/Compliance FDA 21 CFR 177-2600
Color Green
Disposable Single-use
Material Nitrile
Powdered Powdered
Size Large
Thickness (English) 5 mil
Thickness (Metric) 5 mm
Model 92 500 L 585190
Tensile Strength 14 MPa
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less