missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 92-500 Powdered Nitrile Gloves

Catalog No. 18999266B
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18999266B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18999266B Supplier Ansell Supplier No. 9250011080

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powdered gloves made with TNT™ chemical splash resistance technology effectively safeguard hands from exposure to harmful chemicals.

Proven splash resistance against hazardous chemicals. Ansell Touch N Tuff™ Disposable Nitrile Gloves provide a comfortable fit and minimize hand fatigue.

  • Made with TNT™ Chemical Splash Resistance Technology to safeguard from exposure to harmful chemicals.
  • Powdered for moisture absorption and easy donning.
  • Strong proprietary formulation prevents tearing during heavy-duty tasks.
  • Soft and comfortable nitrile material.

Specifications

Certifications/Compliance FDA 21 CFR 177-2600
Accelerator Free No
Applications Laboratory
AQL 1.5
Color Green
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 5.5 mil
Finger Thickness (Metric) 0.14 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.3 mil
Palm Thickness (Metric) 0.11 mm
Powdered Powdered
Product Type Nitrile Gloves
Size Medium
Sterility Non-sterile
Thickness (English) 4.3 mil
Thickness (Metric) 0.11 mm
Glove Cuff Style Beaded
Model 92-500
Series TOUCHNTUFF™ 92-500
Tensile Strength 14 MPa
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less