missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ Solvex™ 37-676 Nitrile Chemical Resistant Gloves

Catalog No. 19120393E
Change view
Click to view available options
Size:
10
11
7
8
9
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19120393E 11
19120393A 7
19120393B 8
19120393C 9
19120393D 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19120393E Supplier Ansell Supplier No. 37676110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitrile gloves deliver optimal results in wet or dry work environments where chemical resistance is crucial.

  • High-performance nitrile compound
  • Protects against abrasions, cuts, snags and punctures
  • Does not contain protein allergens
  • Raised-diamond pattern for a secure grip on wet or slippery materials
  • Form-fitting fingers, contoured palm, and a tapered wrist create a comfortable, ergonomic fit
  • Flock-lining keeps hands cool and dry
  • Ideal for work in meat processing, parts degreasing, machining operations, wiping down parts and machinery, and handling hoses, belts and steel chains
  • Length: 13 in., Thickness: 15 mil

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) EN 1149
Chlorinated Yes
Applications Industrial
Color Green
Finger Thickness (English) 0.0169 in.
Finger Thickness (Metric) 0.43 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 0.0169 in.
Palm Thickness (Metric) 0.43 mm
Product Type Chemical-resistant Gloves
Size 11
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
EN388 Standard EN 388:4101X, EN 388:4101
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Grip Design Reversed Lozenge
Liner Material Cotton Flock
Lining Lined
Model 37-676 11 117725
Series AlphaTec™ Solvex™ 37-676
Length (English) 13 in.
Length (Metric) 33 cm
Show More Show Less