Learn More
Ansell™ Sawyer-Tower™ 66-670 Series Jackets
Catalog No. 19010038
Change view
Click to view available options
Size:
2X-Large
Catalog No. Size
19010038 2X-Large
1 options
Catalog No. 19010038 Supplier Ansell™ Supplier No. 666702XL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Full-coverage chemical protection with added safety of fire resistance
  • Features comfortable Gore™ Chemical Splash Fabric
  • Chemical splash protection
  • Breathable Gore-Tex fabric keeps workers cool in hot situations and warm in cold situations
  • Lightweight, flexible, snag- and tear-resistant, stain-repellent and launderable
  • CPC Nomex provides fire-resistant chemical splash protection
  • Special sizes may be available upon request
  • Flame resistant
  • HRC rating: 2
Specifications
Size 2X-Large
Material CPC Nomex Trilaminate
Color Blue
Arc Flash Rating 2
Resistance Flame, chemical
Product Type Chemical-resistant Jacket
Length (English) 30 in.