missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell ActivArmr™ 80-813 Foam-Coated Flame Resistant Gloves
Flame resistant gloves provide Arc Flash Hazard/Risk Category 2 protection against electrical arc flash hazards. Aramid Fiber shell allows for safe handling of sharp objects.
Supplier: Ansell 8081311080
Description
- Aramid Fiber shell is inherently flame resistant, and allows for safe handling of sharp objects
- Neoprene foam coating enhances grip in dry, wet, or oily conditions
- Arc rating of 12 cal/cm2 meets NFPA 70E Hazard Risk Category 2
- Ergonomic design is comfortable and flexible
Specifications
| ANSI 3 | |
| NFPA 70E Hazard Risk Category 2 | |
| Knit | |
| Green, Black | |
| Specific | |
| Arc Flash Gloves | |
| 2X42C | |
| Knit | |
| ActivArmr™ 80-813 | |
| 13 |
| Level 2 | |
| EN 407: 412110 | |
| Neoprene | |
| ANSI A4 | |
| Aramid Fiber | |
| 8 | |
| Automotive Assembly, Equipment/Machinery Operation, Mining, and Utility Work | |
| ANSI/ISEA 105-16 Level 2 | |
| Wing | |
| 8.86 to 10.24 in. |