missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-813 Foam-Coated Flame Resistant Gloves

Catalog No. 19823450
Change view
Click to view available options
Size:
10
11
6
8
9
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19823450 6
19823452 8
19823453 9
19823454 10
19823455 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19823450 Supplier Ansell Supplier No. 8081311060

Flame resistant gloves provide Arc Flash Hazard/Risk Category 2 protection against electrical arc flash hazards. Aramid Fiber shell allows for safe handling of sharp objects.

  • Aramid Fiber shell is inherently flame resistant, and allows for safe handling of sharp objects
  • Neoprene foam coating enhances grip in dry, wet, or oily conditions
  • Arc rating of 12 cal/cm2 meets NFPA 70E Hazard Risk Category 2
  • Ergonomic design is comfortable and flexible

Specifications

ANSI Abrasion Rating ANSI 3
Arc Flash Level 2
Certifications/Compliance NFPA 70E Hazard Risk Category 2
Certifications/Compliance (Europe) EN 407: 412110
Construction Knit
Coating Material Neoprene
Color Green, Black
Cut Resistance Rating ANSI A4
Hand Specific
Material Aramid Fiber
Product Type Arc Flash Gloves
Size 6
EN388 Standard 2X42C
For Use With (Application) Automotive Assembly, Equipment/Machinery Operation, Mining, and Utility Work
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series ActivArmr™ 80-813
Thumb Style Wing
Yarn Gauge 13
Length (English) 8.86 to 10.24 in.
Length (Metric) 22.5 to 26 cm
Show More Show Less