missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICRO-TOUCH™ Nitrile E.P.™ Non-Sterile Gloves

Catalog No. p-4840055
Change view
Click to view available options
Size:
X-Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17200843 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17200843 Supplier Ansell Supplier No. 6034050
Only null left

Nitrile, powder free examination glove with an extended cuff provides extra protection when healthcare providers are exposed to chemotherapy agents, bodily fluids or blood-borne pathogens in high-risk clinical settings.

  • Non-sterile glove with extended cuff
  • Ideal for healthcare providers in high-risk clinical settings
  • Chemical and puncture resistant
  • Textured for better grip during wet and dry procedures
  • Tested for use with chemotherapy drugs

Specifications

Certifications/Compliance ASTM D6319, ASTM D6978, FDA21 CFR 177-2600, ISO 13485, ISO 9001, ISO 10993-10:2002
Certifications/Compliance (Europe) EN 374:2003, EN 420:2003 + A1:2009
Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Material Nitrile
Size X-Small
Model 6034050
Length (English) 11.6 in.
Length (Metric) 29.4 cm
Show More Show Less