missing translation for 'onlineSavingsMsg'
Learn More

Ansell Hynit™ 32-125 Nitrile Coated Liquid Repellent Gloves

Catalog No. p-144661
Change view
Click to view available options
Size:
7
7.5
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19019342 7
19019344 7.5
19019330 8
19019331 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19019342 Supplier Ansell Supplier No. 3212511070
Only null left

Special nitrile-coating formulation is flexible while also repelling liquid, oil, and dirt. Gloves are compliant with FDA Food Handling Requirements.

  • Liquid-repellent impregnation repels oil, grease, and dirt
  • Nitrile coating is flexible for optimum dexterity
  • Compliant with FDA food handling requirements
  • Resistant to abrasions

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Copper, Polyester
Product Type Dip-coated Gloves
Size 7
For Use With (Application) Automotive Assembly, Equipment Operation, Metal Fabrication, and Mining
Glove Cuff Style Open
Grip Design Smooth
Liner Material Cotton/Polyester Blend
Lining Lined
Packaging Type Polybag
Puncture Rating ANSI/ISEA 105-16 Level 2
Series 32-125
Thumb Style Wing
Show More Show Less