missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 47-402 Nitrile Coated Medium Duty Gloves

Catalog No. 2870926
Change view
Click to view available options
Size:
10
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
2870926 9
19019880 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2870926 Supplier Ansell Supplier No. 4740211090

Nitrile coating is abrasion and oil-resistant to enhance grip capabilities in dry or oily conditions. Gloves feature interlock knit lining, generous sizing, and a contoured design for increased comfort.

  • ActivArmr™ gloves combine the comfort of cotton gloves with the abrasion resistance and oil repellence of a nitrile-coated glove
  • Interlock knit lining, generous sizing, and contoured designs increase comfort
  • Special nitrile formulation provides mechanical resistance and oil repellency
  • Designed to conform to the hand's natural shape for long wear periods
  • Ensure cleanliness of metal sheets in the steel and motor industries
  • Ideal for a variety of medium-duty job tasks

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Full Glove
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 9
For Use With (Application) Automotive Repairs, Heavy Equipment Operation, Metal Fabrication, and Chemical Handling
Glove Cuff Style Knit
Grip Design Smooth
Packaging Type Polybag
Puncture Rating ANSI/ISEA 105-16 Level 2
Series 47-402
Thumb Style Wing
Show More Show Less