missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-727 Medium-Duty, 15 Gauge Cut Protection Gloves, INTERCEPT™ Technology

Abrasion-resistant gloves feature a polyurethane coating for increased comfort, as well as a secure grip on dry to slightly oily pieces for secure handling in a wide range of environments.

Supplier:  Ansell 11727100

 View more versions of this product

Catalog No. 19-010-110

Only null left
Explore available promotions

Description

Description

  • Lightweight, easy to wear during long working hours
  • Liner features optimized Lycra™ content for cooling effect on the skin
  • Silicone-free polyurethane (PU) palm coating provides enhanced grip in light oil environments for secure handling
  • Gray coating hides dirt in environments with dirt exposure
  • ANSI cut level 2
  • EN Abrasion Level 4 (ANSI Level 6)
  • Available in sizes 6 through 11
Applications:
  • Assembling metal parts and components
  • Assembling white goods
  • Cutting of small dry or lightly oiled parts and maintenance
  • General maintenance
  • Glass manufacturing
  • Handling of sharp-edged objects
  • Metal fabrication
  • Plastics injection and moulding
  • Pre-assembling
Specifications

Specifications

ANSI 6
Knit
Gray
Specific
HPPE, Nylon, Spandex, Polyethylene
10
Automotive, machinery and equipment, metal fabrication, chemical, automotive aftermarket, EMS, utilities, warehousing
Knit
HPPE, Nylon, Spandex
11-727 10 163840
ANSI/ISEA 105-16 Level 3
Wing
19.5 cm
CE, EN 420:2003 + A1:2009; Category II
Polyurethane
ANSI A2
Yes
Cut-resistant Gloves
EN 388:4342
Unisex
Rough
Lined
Case
HyFlex™ 11-727
7.6 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions