missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-920 Nitrile Coated Oil Repellent Gloves

Catalog No. p-3802305
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19148500 X-Small
19148501 Small
19148502 Medium
19148503 Large
19148504 X-Large
19148505 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19148500 Supplier Ansell Supplier No. 1192011060
Only null left

Seamless knit gloves feature ANSELL GRIP™ Technology which utilizes microscopic channels in the glove coating to displace oil, enhancing grip capabilities when working with oily parts.

  • High performance grip and oil repellence when working in oily environments
  • ANSELL GRIP™ Technology contains microscopic channels in the glove coating to displace oil
  • RIPEL™ Liquid Repellence Technology creates a liquid impermeable layer to prevent oil or lubricant from making contact with the wearer's skin
  • Seamless knit design provides a comfortable fit with maximum dexterity

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
For Use With (Application) Automotive Assembly, Equipment Operation, Machinery, and Metal Fabrication
Glove Cuff Style Knit
Series 11-920
Yarn Gauge 15
Show More Show Less