missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-920 Nitrile Coated Oil Repellent Gloves

Catalog No. 19148500
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19148500 X-Small
19148501 Small
19148502 Medium
19148503 Large
19148504 X-Large
19148505 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19148500 Supplier Ansell Supplier No. 1192011060

Seamless knit gloves feature ANSELL GRIP™ Technology which utilizes microscopic channels in the glove coating to displace oil, enhancing grip capabilities when working with oily parts.

  • High performance grip and oil repellence when working in oily environments
  • ANSELL GRIP™ Technology contains microscopic channels in the glove coating to displace oil
  • RIPEL™ Liquid Repellence Technology creates a liquid impermeable layer to prevent oil or lubricant from making contact with the wearer's skin
  • Seamless knit design provides a comfortable fit with maximum dexterity

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI/ISEA 105-2024 (A1), CE Category II, EN 388:2016 (3131A), EN ISO 21420:2020, REACH
Certifications/Compliance (Europe) CE Category II, EN 388:2016 (3131A), EN ISO 21420:2020, REACH
Chemotherapeutic Rated No
Coated Area Palm
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
For Use With (Application) Automotive Assembly, Equipment Operation, Machinery, and Metal Fabrication
Glove Cuff Style Knit
Grip Design Ansell Grip Technology
Liner Material Nylon
Resistant To Oil, Lubricants
Series 11-920
Yarn Gauge 15
Length (English) 8.66 to 10.63 in.
Length (Metric) 220 to 270 mm
Product Line HyFlex
Show More Show Less