missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-900 Nitrile Coated Oil Repellent Gloves

Catalog No. p-3348224
Change view
Click to view available options
Size:
10
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296355659 7
296355661 8
296355662 9
296355663 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296355659 Supplier Ansell Supplier No. 1190011060
Only null left

Industrial work gloves designed for light mechanical protection, particularly in moderately oily manufacturing or maintenance environments.

  • Nitrile coating provides light mechanical protection and dexterity
  • Designed for use in moderately oily manufacturing and maintenance environments
  • Combines protection with enhanced grip and abrasion resistance

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Chlorinated No
Coated Area Palm, Fingers
Construction Knit
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 7
For Use With (Application) Assembly, Equipment Operation, and Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Series 11-900
Yarn Gauge 13
Show More Show Less