missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-900 Nitrile Coated Oil Repellent Gloves

Catalog No. 296355662
Change view
Click to view available options
Size:
10
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296355662 9
296355659 7
296355661 8
296355663 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296355662 Supplier Ansell Supplier No. 1190011090

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Industrial work gloves designed for light mechanical protection, particularly in moderately oily manufacturing or maintenance environments.

  • Nitrile coating provides light mechanical protection and dexterity
  • Designed for use in moderately oily manufacturing and maintenance environments
  • Combines protection with enhanced grip and abrasion resistance

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI 4, EN 388: 4121A, EN 420:2003 + A1:2009, Category II, REACH
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Chlorinated No
Coated Area Palm, Fingers
Construction Knit
Coating Material Nitrile
Color White, Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 9
For Use With (Application) Assembly, Equipment Operation, and Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Liner Material Nylon
Series 11-900
Yarn Gauge 13
Length (English) 7.87 to 10.04 in.
Length (Metric) 200 to 255 mm
Product Line HyFlex
Show More Show Less